missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PR10A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA598368
This item is not returnable.
View return policy
Description
Involved in the biosynthesis of the common precursor of all benzylisoquinoline alkaloids such as morphine, sanguinarine, codeine or berberine. Condenses dopamine and pyruvic acid or 4-hydroxyphenylpyruvate. [Uniprot]
Specifications
PR10A | |
Polyclonal | |
Unconjugated | |
CjPR10A, Pathogenesis related protein 10A, S-norcoclaurine synthase 2 | |
Recombinant Coptis japonica S-norcoclaurine synthase 2 protein (20-196AA) | |
100 μg | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Plant | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PR10A Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction