missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium foil, 0.1mm (0.004in) thick, 99% (REO), Thermo Scientific Chemicals

Product Code. 11480380
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm

Praseodymium optical properties for use in amplification of telecommunication systems, including as a doping agent in fluoride fibers. It is also used in the scintillator for medical CAT scans. Used as additive for special steel, non-ferric alloy, hydrogen storage alloy, also reducer for making other rare Earth metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-10-0
Molecular Formula Pr
Molecular Weight (g/mol) 140.91
MDL Number MFCD00011174
InChI Key PUDIUYLPXJFUGB-UHFFFAOYSA-N
Synonym unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
PubChem CID 23942
ChEBI CHEBI:49828
IUPAC Name praseodymium
SMILES [Pr]

Specifications

UN Number UN3178
Quantity 50 x 50 mm
Solubility Information Reacts with water.
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.42g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Praseodymium foil, 0.1mm (0.004 in.) thick

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.