missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11362568
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Praseodymium Acetate is used to make color glasses and enamels; when mixed with certain other materials, Praseodymium produces an intense clean yellow color in glass. The other main application is used to manufacturing pigment and ink color.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334869-74-8
Synonym n-2-chlorophenyl-2-trifluoromethyl benzamide, cbmicro_015862
PubChem CID 875825

Specifications

Physical Form Crystalline Aggregates
Quantity 25 g
Molecular Formula C14H9ClF3NO
MDL Number MFCD02219578
Solubility Information Soluble in water,moderately soluble in strong mineral acids.
InChI Key CBVKYRGTGNCYLK-UHFFFAOYSA-N
SMILES FC(F)(F)C1=CC=CC=C1C(=O)NC1=CC=CC=C1Cl
IUPAC Name N-(2-chlorophenyl)-2-(trifluoromethyl)benzamide
Molecular Weight (g/mol) 299.68
Formula Weight 318.03 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Praseodymium(III) acetate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'