missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11388557
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Praseodymium(III) bromide has some of the common uses such as in aircraft engines, motion picture industry, coloring in glasses and enamels, color cubic zirconia to simulate peridot, welder and glass blower goggles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13536-53-3
MDL Number MFCD00049762

Specifications

Melting Point 691°C
Physical Form Powder
Quantity 2 g
Solubility Information It is soluble in water.
Formula Weight 380.63
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Praseodymium(III) bromide, Ultra dry

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'