missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) carbonate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11352278
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14948-62-0
Molecular Formula C3H16O17Pr2
Molecular Weight (g/mol) 605.959
MDL Number MFCD00211307
InChI Key SEHQEMTYQGRHIC-UHFFFAOYSA-H
Synonym praseodymium carbonate octahydrate, 3co3.2pr.8h2o, praseodymium carbonate, praseodymium carbonate-water 2/3/8, dipraseodymium 3+ ion octahydrate trico3, praseodymium iii carbonate octahydrate, reacton, dipraseodymium 3+ octahydrate tricarbonate
PubChem CID 25022096
IUPAC Name praseodymium(3+);tricarbonate;octahydrate
SMILES C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.O.O.O.O.[Pr+3].[Pr+3]

Specifications

Physical Form Crystalline
Quantity 5 g
Formula Weight 605.97 (461.85 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Assay (REO)
Chemical Name or Material Praseodymium(III) carbonate octahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'