missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

80.90 € - 273.36 €

Chemical Identifiers

CAS 19423-77-9
Molecular Formula Cl3Pr
Molecular Weight (g/mol) 247.26
MDL Number MFCD00149828
InChI Key LHBNLZDGIPPZLL-UHFFFAOYSA-K
IUPAC Name praseodymium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Pr+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11316298
View Documents
Thermo Scientific Alfa Aesar
011237.14
25 g
80.90 €
25g
Please to purchase this item. Need a web account? Register with us today!
11306298
View Documents
Thermo Scientific Alfa Aesar
011237.22
100 g
273.36 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aqueous solutions of praseodymium(III) chloride can be used to prepare insoluble praseodymium(III) compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19423-77-9
247.26
LHBNLZDGIPPZLL-UHFFFAOYSA-K
[Cl-].[Cl-].[Cl-].[Pr+3]
Cl3Pr
MFCD00149828
praseodymium(3+) trichloride

Specifications

115°C
Crystalline Aggregates
MFCD00149828
Soluble in water and alcohol
[Cl-].[Cl-].[Cl-].[Pr+3]
247.26
99.9%
Odorless
Praseodymium(III) chloride hydrate
19423-77-9
Cl3Pr
14,7712
LHBNLZDGIPPZLL-UHFFFAOYSA-K
praseodymium(3+) trichloride
247.27 (Anhydrous)
Reacton™
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'einecsNumber' : 233-794-4

missing translation for 'rtecsNumber' : TU0175000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Praseodymium(III) chloride hydrate, REacton, 99.9% (REO)

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.

Supplemental information
  • MIXTURE LIST-Contains : Praseodymium(III) chloride hydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.