Learn More
Praseodymium(III) chloride, ultra dry, 99.95% (REO), Thermo Scientific Chemicals
55.00 € - 358.79 €
Chemical Identifiers
CAS | 10361-79-2 |
---|---|
MDL Number | MFCD00011176 |
Synonym | acmc-20ajts, praseodymium 3+ ion trichloride, praseodymium chloride, anhydrous, praseodymium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 5g |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15439105
|
Thermo Scientific Alfa Aesar
H37815.03 |
1 g | N/A | ||||||
15449105
|
Thermo Scientific Alfa Aesar
H37815.06 |
5 g |
146.73 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15459105
|
Thermo Scientific Alfa Aesar
H37815.14 |
25 g |
358.79 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
10361-79-2 | |
MFCD00011176 | |
acmc-20ajts, praseodymium 3+ ion trichloride, praseodymium chloride, anhydrous, praseodymium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 5g |
Specifications
1700°C | |
10361-79-2 | |
PrCl3 | |
acmc-20ajts, praseodymium 3+ ion trichloride, praseodymium chloride, anhydrous, praseodymium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 5g | |
99.95% | |
Ampule | |
4.02 g/mL | |
Praseodymium(III) chloride, Ultra dry |
786°C | |
Powder | |
MFCD00011176 | |
247.27 | |
Odorless | |
Hygroscopic | |
(REO) |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'rtecsNumber' : TU0175000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.