missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) iodide, ultra dry, 99.9+% (REO), Thermo Scientific Chemicals

Product Code. 11318168
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13813-23-5
Molecular Formula I3Pr
Molecular Weight (g/mol) 521.621
MDL Number MFCD00049513
InChI Key PVEVRIVGNKNWML-UHFFFAOYSA-K
Synonym praseodymium triiodide, praseodymium iodide pri3, pri3, prazodymium iodide, praseodymium iii iodide, ultra dry
PubChem CID 83744
IUPAC Name triiodopraseodymium
SMILES I[Pr](I)I

Specifications

Melting Point 737°C
Physical Form Crystals or Needles
Quantity 1 g
Linear Formula PrI3
Solubility Information Soluble in water.
Formula Weight 521.62
Percent Purity ≥99.9%
Packaging Ampule
Sensitivity Hygroscopic; Light sensitive
Assay (REO)
Chemical Name or Material Praseodymium(III) iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Praseodymium(III) iodide, ultra dry, 99.9+% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.