Learn More
Praseodymium(III, IV) oxide, 99% (REO), Thermo Scientific Chemicals
Description
Used in ceramic structures. Have a wide range of applications from producing clay bowls to advanced electronics. Praseodymium oxide is also useful in light weight structural components in aerospace and electrochemical applications such as fuel cells due to its ionic conductivity. Component of didymium glass which is a colorant for welder's goggles, also as important additive of Praseodymium yellow pigments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 2500°C |
Physical Form | Powder |
Quantity | 5 kg |
Solubility Information | Insoluble in water and only soluble in strong acid. |
Formula Weight | 1021.43 |
Percent Purity | 99% |
Assay | (REO) |
Chemical Name or Material | Praseodymium (III, IV) oxide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.