Learn More
Praseodymium(III, IV) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
296.00 € - 1051.00 €
Chemical Identifiers
CAS | 12037-29-5 |
---|---|
Molecular Formula | O11Pr6 |
Molecular Weight (g/mol) | 1021.44 |
MDL Number | MFCD00011178 |
InChI Key | HPZIIFVSYNLWNX-UHFFFAOYSA-N |
IUPAC Name | hexapraseodymium(3+) undecaoxidandiide |
SMILES | [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386288
|
Thermo Scientific Alfa Aesar
011235.14 |
25 g |
296.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396288
|
Thermo Scientific Alfa Aesar
011235.22 |
100 g |
1051.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Praseodymium(III, IV) oxide is used in pigment in glass, optic and ceramic industries. Praseodymium-doped glass, called didymium glass is used in welding, blacksmithing, and glass-blowing goggles due to its blocking property of infrared radiation. It is employed in the solid state synthesis of praseodymium molybdenum oxide, which is used as a semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12037-29-5 | |
1021.44 | |
HPZIIFVSYNLWNX-UHFFFAOYSA-N | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3] |
O11Pr6 | |
MFCD00011178 | |
hexapraseodymium(3+) undecaoxidandiide |
Specifications
2500°C | |
Black | |
99.99% (REO) | |
MFCD00011178 | |
HPZIIFVSYNLWNX-UHFFFAOYSA-N | |
hexapraseodymium(3+) undecaoxidandiide | |
1021.43 | |
Praseodymium(III, IV) oxide |
12037-29-5 | |
Powder | |
O11Pr6 | |
Insoluble in water. Moderately soluble in strong mineral acids. | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3] | |
1021.44 | |
Black powder |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 234-857-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only