missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11346298
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Praseodymium(III) nitrate hexahydrate is used in electrical components, optical glasses and photo-optical materials. Also find use as catalysts and laboratory reagent. In addition, it is used in the solvothermal process for formation of doped lanthanide oxysulfide. It is a precursor for the electrodeposition of praseodymium oxide for electrochemical applications. It is a starting material for the preparation of ceramic pigments, metal-organic frameworks and mixed conductors. It finds utility in the synthesis of other praseodymium compounds and complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15878-77-0
Molecular Formula H12N3O15Pr
Molecular Weight (g/mol) 435.01
MDL Number MFCD00149826
InChI Key LXXCECZPOWZKLC-UHFFFAOYSA-N
Synonym praseodymium iii nitrate hexahydrate, praseodymium iii nitrate, hexahydrate 1:3:6, praseodymium nitrate hexahydrate, nitric acid, praseodymium 3+ salt, hexahydrate, praseodymium 3+ hexahydrate trinitrate, ksc529q4r, praseodymium trinitrate hexahydrate, praseodymium nitrate hydrate 1:3:6, praseodymium nitrate,, praseodymium 3+ trinitrate hexahydrate
PubChem CID 204170
IUPAC Name praseodymium(3+);trinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 25 g
UN Number UN1477
Solubility Information Soluble in water and alcohol.
Formula Weight 435.01 (326.92 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Praseodymium(III) nitrate hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Praseodymium(III) nitrate hexahydrate, REacton
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contains : Praseodymium(III) nitrate hexahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'