Learn More
Praseodymium(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Description
Praseodymium(III) nitrate hexahydrate is used in electrical components, optical glasses and photo-optical materials. Also find use as catalysts and laboratory reagent. In addition, it is used in the solvothermal process for formation of doped lanthanide oxysulfide. It is a precursor for the electrodeposition of praseodymium oxide for electrochemical applications. It is a starting material for the preparation of ceramic pigments, metal-organic frameworks and mixed conductors. It finds utility in the synthesis of other praseodymium compounds and complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Physical Form | Crystalline Aggregates |
Quantity | 100 g |
UN Number | UN1477 |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 435.01 (326.92 Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Odor | Odorless |
Sensitivity | Hygroscopic |
Assay | (REO) |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.