Learn More
Praseodymium(III) nitrate hydrate, 99.9% (REO)
Beskrivelse
Praseodymium(III) nitrate hydrate is used in fluorescent display tubes and phosphors. It is also used in the ultrasonic synthesis of phase-pure praseodymium molybdate. It plays a major role in the solvothermal process for formation of doped lanthanide oxysulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| Color | Green |
| Physical Form | Crystalline |
| Quantity | 250 g |
| Assay Percent Range | 99.9% |
| Solubility Information | Soluble in water,anhydrous amine,alcohol,ether,polar solvents and acetonitrile. |
| IUPAC Name | praseodymium(3+);trinitrate;hexahydrate |
| Formula Weight | 326.92 (anhydrous) |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Appearance | Green crystals |
| Vis mere |
RUO – Research Use Only
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.