missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11317647
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Praseodymium(III) nitrate hydrate is used in fluorescent display tubes and phosphors. It is also used in the ultrasonic synthesis of phase-pure praseodymium molybdate. It plays a major role in the solvothermal process for formation of doped lanthanide oxysulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15878-77-0
Molecular Formula H12N3O15Pr
Molecular Weight (g/mol) 435.01
MDL Number MFCD00011175
InChI Key LXXCECZPOWZKLC-UHFFFAOYSA-N
Synonym praseodymium iii nitrate hexahydrate, praseodymium iii nitrate, hexahydrate 1:3:6, praseodymium nitrate hexahydrate, nitric acid, praseodymium 3+ salt, hexahydrate, praseodymium 3+ hexahydrate trinitrate, ksc529q4r, praseodymium trinitrate hexahydrate, praseodymium nitrate hydrate 1:3:6, praseodymium nitrate,, praseodymium 3+ trinitrate hexahydrate
PubChem CID 204170
SMILES O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Green
Physical Form Crystalline
Quantity 10 g
Assay Percent Range 99.9%
Solubility Information Soluble in water,anhydrous amine,alcohol,ether,polar solvents and acetonitrile.
IUPAC Name praseodymium(3+) hexahydrate trinitrate
Formula Weight 326.92 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Green crystals
Chemical Name or Material Praseodymium(III) nitrate hydrate, Rare Earth Oxides Content
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'