missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific™

94.00 € - 278.00 €

Chemical Identifiers

CAS 13498-07-2
Molecular Formula Cl3O12Pr
Molecular Weight (g/mol) 439.246
MDL Number MFCD00243288
InChI Key OPAITELCMCAYEB-UHFFFAOYSA-K
Synonym praseodymium 3+ perchlorate, praseodymium iii perchlorate, acmc-20ajtn, praseodymium perchlorate, praseodymium iii perchlorate solution, trihyperchloric acid praseodymium iii salt
PubChem CID 13783608
IUPAC Name praseodymium(3+);triperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Pr+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11450291
View Documents
Thermo Scientific Alfa Aesar
040569.NC
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11460291
View Documents
Thermo Scientific Alfa Aesar
040569.NE
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13498-07-2
439.246
OPAITELCMCAYEB-UHFFFAOYSA-K
13783608
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Pr+3]
Cl3O12Pr
MFCD00243288
praseodymium 3+ perchlorate, praseodymium iii perchlorate, acmc-20ajtn, praseodymium perchlorate, praseodymium iii perchlorate solution, trihyperchloric acid praseodymium iii salt
praseodymium(3+);triperchlorate

Specifications

13498-07-2
Cl3O12Pr
UN3211
OPAITELCMCAYEB-UHFFFAOYSA-K
praseodymium(3+);triperchlorate
13783608
Reagent
Odorless
Liquid
MFCD00243288
praseodymium 3+ perchlorate, praseodymium iii perchlorate, acmc-20ajtn, praseodymium perchlorate, praseodymium iii perchlorate solution, trihyperchloric acid praseodymium iii salt
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Pr+3]
439.246
439.26
50% w/w aq. soln.
Praseodymium(III) perchlorate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H271-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.

missing translation for 'einecsNumber' : 236-822-3

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.