missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11342278
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13510-41-3
Molecular Formula H16O20Pr2S3
Molecular Weight (g/mol) 714.103
MDL Number MFCD00149832
InChI Key NIJXLQIGVGZNKP-UHFFFAOYSA-H
Synonym praseodymium sulfate-water 2/3/8, 3so4.2pr.8h2o, praseodymium sulfate, dipraseodymium 3+ ion octahydrate trisulfate, praseodymium iii sulfate octahydrate, reacton, dipraseodymium 3+ octahydrate trisulfate
PubChem CID 25022097
IUPAC Name praseodymium(3+);trisulfate;octahydrate
SMILES O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Pr+3].[Pr+3]

Specifications

Physical Form Powder
Quantity 100 g
Merck Index 14,7712
Formula Weight 714.12 (570.00 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Assay (REO)
Chemical Name or Material Praseodymium(III) sulfate octahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'