missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium ingot, 99% (REO), Thermo Scientific Chemicals

Product Code. 11346507
Click to view available options
Quantity:
10 g
50 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-10-0
Molecular Formula Pr
Molecular Weight (g/mol) 140.91
MDL Number MFCD00011174
InChI Key PUDIUYLPXJFUGB-UHFFFAOYSA-N
Synonym unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
PubChem CID 23942
ChEBI CHEBI:49828
IUPAC Name praseodymium
SMILES [Pr]

Specifications

Color Gray to Silver
Appearance Gray to silver ingot
Density 6.773 g/cm3
Assay Percent Range 99.9%
Quantity 50 g
Odor Odorless
Sensitivity Air and moisture sensitive
Chemical Name or Material Praseodymium, ingot
compliance-icons
Product Identifier
  • Praseodymium
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H228-Flammable solid.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Praseodymium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.