missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium powder, -40 mesh, 99.5% (REO)

Product Code. 15411977
Click to view available options
Quantity:
5 g
Packaging:
Ampule
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15411977

missing translation for 'mfr': Thermo Scientific Alfa Aesar 044183.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-10-0 | Pr | 140.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Specifications

Color Gray to Silver
CAS 7440-10-0
Quantity 5 g
Mesh Size −40 Mesh
Odor Odorless
Packaging Ampule
Sensitivity Air and moisture sensitive
Form Powder
Molecular Formula Pr
MDL Number MFCD00011174
UN Number UN3089
Synonym unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
InChI Key PUDIUYLPXJFUGB-UHFFFAOYSA-N
SMILES [Pr]
IUPAC Name praseodymium
Molecular Weight (g/mol) 140.91
PubChem CID 23942
ChEBI CHEBI:49828
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Praseodymium powder
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.