missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Prasugrel, 96%, Thermo Scientific Chemicals Product Code.: 15698735

Thermo Scientific Chemicals Prasugrel, 96%, Thermo Scientific Chemicals

Product Code. 15698735
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15698735

Brand: Thermo Scientific Chemicals 461482500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150322-43-3
Molecular Formula C20H20FNO3S
Molecular Weight (g/mol) 373.44
InChI Key DTGLZDAWLRGWQN-UHFFFAOYSA-N
Synonym prasugrel, effient, efient, prasugrel effient, prasita, 5-2-cyclopropyl-1-2-fluorophenyl-2-oxoethyl-4,5,6,7-tetrahydrothieno 3,2-c pyridin-2-yl acetate, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl ethanone, effient tn, ethanone, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenylShow MoreShow Less
PubChem CID 6918456
ChEBI CHEBI:87723
IUPAC Name [5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-6,7-dihydro-4H-thieno[3,2-c]pyridin-2-yl] acetate
SMILES CC(=O)OC1=CC2=C(S1)CCN(C2)C(C3=CC=CC=C3F)C(=O)C4CC4

Specifications

Melting Point 110.0°C to 121.0°C
Ignition Residue 0.1% max
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Formula Weight 373.44
Percent Purity 98%
Chemical Name or Material Prasugrel

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458955 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog Number461482500,461480010
Lot NumberA0375333 Certificate TypeCertificate of Analysis Date14/09/2021 Catalog Number461482500,461480010
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Prasugrel, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.