Learn More
Brady™ Pre-cut Blank Labels for BBP3X, S3XXX, and i3300 Printers
The material of these labels is ideal for indoor and outdoor facility labels.
Brand:  Brady™ 120931
 View more versions of this product
Product Code. 15981001
  • 203.00 € / Pack of 300
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels
  • Vinyl material
Specifications
Specifications
Not Autoclavable
White
Vinyl (B-7569)
Non-sterile
BBP™30, BBP™31, BBP™33, BBP™35 Multicolor, BBP™37 Multicolor, BradyPrinter i3300, S3000, S3100, and i5300 Industrial Label Printers
300 Labels
-20°C to +75°C
82.6 mm
75 mm
Permanent Acrylic
Gloss
Thermal Transfer
Facility Identification, Hazard Communication Identification, Electrical Labeling
1
Rectangle
0.102 mm
57 mm