Learn More
Ambion™ Pre-miR™ miRNA Precursor
Pre-miR™ miRNA Precursor
325.00 €
Specifications
Product Line Ambion™, Pre-miR™
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10782307
View Documents
Ambion™
AM17100
5 nmol
325.00 €
Each
Description
Description
miRNA Precursor Molecules are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs.
Specifications
Specifications
Ambion™, Pre-miR™