Learn More
Bochem™ Precision Forceps
Manufactured from 18/10 stainless steel
96.90 €
Specifications
Tip Style Sharp
No. per Pack 1 Pack
Material Stainless Steel
Length (Metric) 120 mm
Product Type Forceps, Precision
View More Specs
Products 1
Product Code Brand Length (Metric) Tip Style Price Quantity & Availability  
Product Code Brand Length (Metric) Tip Style Price Quantity & Availability  
11895151
View Documents
Bochem™
1640
120 mm Sharp
96.90 €
Pack of 10
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
Description
Description
  • Points without ridges
  • Self-tensing
Specifications
Specifications
Sharp
Stainless Steel
Forceps, Precision
1 Pack
120 mm
Forceps