missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gestigkeit Harry Precision Hotplate
Precision hotplates, PZ-series for continuous operation. Provide extremely accurate, uniform temperatures, even in plate corners and on edges.
Brand: Gestigkeit Harry PZ44EB/230
This item is not returnable.
View return policy
Description
- For continuous operation.
- Provide extremely accurate, uniform temperatures, even in plate corners and on edges.
- Polished anodised aluminum heating surface.
- Microprocessor-controlled temperature controller with temperature setting up to 99.9 in 0.1°C steps, over 99.9 in 1°C steps. Actual temperature displayed continuously.
- With separate power controller for performance adjustment to the heating requirements from 10 to 100%.
- Built-in relay allows direct connection of electronic contact thermometers.
- Adjustable temperature monitor from 50 to 300°C (with PZ 28-1, 30 to 110°C) to prevent excess temperatures.
Specifications
440 x 290 x 190 mm | |
Digital | |
Microprocessor Control | |
3300 W |
Digital Hotplate | |
230 V | |
20°C to 450°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction