Learn More
Thermo Scientific™ Precision™ Shaking Water Baths
Get shaking water baths that support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culture and genetic sequencing.
Brand: Thermo Scientific™ TSSWB27
3226.95 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Description
Benefit from Thermo Scientific™ Precision™ shaking water baths, which support a range of sensitive life science and QA/QC applications – from warming fragile reagents to tissue culturing and genetics sequencing. Easily clean and maintain your bath with new coil-free interior. The Precision shallow form shaking bath has a removable tray that allows for use with smaller sample containers. Precision Dubnoff shaking water baths are designed specifically for applications that require your samples to be incubated in a controlled atmosphere
- Easily clean and maintain bath with coil-free design
- Optimize scheduling with auto-on and auto-off timers
- Help prevent bath overheating and damage with low-fluid protection
- Conveniently save commonly used settings with four temperature and shaking speed presets
- Help protect your work with audible alarms
- Easily operate and monitor with icon-based graphical display
- Accommodate taller labware with new hinged lid and extended height
- Features next-generation shaker motor
Specifications
304 Stainless Steel | |
27L Shaking Water Bath | |
UL, CE | |
Monochrome LCD | |
Stainless Steel Gable Cover and Rubber Duck | |
Precision SWB 27 | |
4 | |
±0.05°C | |
Global Voltage 100/115 V 200/230 V 50/60 Hz |
Digital | |
27 L | |
Ambient +5° to 100°C | |
1500 w | |
Epoxy Powder-coated Cold Rolled Steel | |
N5-20. (Requires a 20A receptacle.) | |
±0.1°C | |
15.5 x 36.9 x 9.8 in. (394 x 938 x 249 mm) | |
11.2-13.1 / 5.4-6.3 (120V/230V) |