missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Precision™ Shaking Water Baths

Get shaking water baths that support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culture and genetic sequencing.

Brand:  Thermo Scientific™ TSDUB15

 View more versions of this product

Product Code. 15395867

  • 3507.45 € / Each
Estimated Shipment: 16-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Benefit from Thermo Scientific™ Precision™ shaking water baths, which support a range of sensitive life science and QA/QC applications – from warming fragile reagents to tissue culturing and genetics sequencing. Easily clean and maintain your bath with new coil-free interior. The Precision shallow form shaking bath has a removable tray that allows for use with smaller sample containers. Precision Dubnoff shaking water baths are designed specifically for applications that require your samples to be incubated in a controlled atmosphere

  • Easily clean and maintain bath with coil-free design
  • Optimize scheduling with auto-on and auto-off timers
  • Help prevent bath overheating and damage with low-fluid protection
  • Conveniently save commonly used settings with four temperature and shaking speed presets
  • Help protect your work with audible alarms
  • Easily operate and monitor with icon-based graphical display
  • Accommodate taller labware with new hinged lid and extended height
  • Features next-generation shaker motor
TRUSTED_SUSTAINABILITY
Specifications

Specifications

304 Stainless Steel
15L Dubnoff Shaking Water Bath
UL, CE
Monochrome LCD
Large and Small Gassing Hood and Rubber Duck
Precision DUB 15
±0.1°C
25 x 15.6 x 9.9 in. (636 x 396 x 250 mm)
9.0-10.5 / 4.3-5.0 (120V/230V)
Digital
15 L
Ambient +5° to 100°C
1200 w
Epoxy Powder-coated Cold Rolled Steel
4
±0.05°C
Global Voltage 100/115 V 200/230 V 50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.