Learn More
Fisherbrand™ Prefiltration Kit
Includes three stages of prefiltration. Fisher Scientific™ Prefiltration Kit protects RO membranes by removing organics, chlorine, and particles from tap water prior to RO filtration.
Brand:  Fisherbrand™ RW10311KT
Product Code. 15439759
  • 264.00 € / Each
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Activated carbon
  • Anti-scaling
  • Depth filtration
Specifications
Specifications
3-Stage, 254mm (10 in.)
Accu500
Water purification