Learn More
Lonza™ PrEGM™ Prostate Epithelial Cell Growth Medium SingleQuots™ Supplements and Growth Factors
Supplements and growth factors kit for use with prostate epithelial cell basal medium.
Brand:  Lonza™ CC-4177
Product Code. 11695500
  • 509.09 € / Each
Explore more special offers
Description
Description
  • Separately sold SingleQuot™ kit for supplementing basal medium.
  • Part of the full PrEGM™ -2 Prostate Epithelial Cell Growth Medium BulletKit™ (catalog number CC-3166), which consists of basal medium and supplements packaged together.
  • Includes the following:
    • 1 x Orange Cap Vial with BPE 2.00 mL
    • 1 x Amber Vial with Triiodothyronine 0.50 mL
    • 1 x Lilac Cap Vial with Insulin 0.50 mL
    • 1 x Green Cap Vial with hEGF 0.50 mL
    • 1 x Natural Cap Vial with Hydrocortisone 0.50 mL
    • 1 x Natural Cap Vial with Transferrin 0.50 mL
    • 1 x Amber Vial with Epinephrine 0.50 mL
    • 1 x Red Cap Vial with GA-1000 0.50 mL
    • 1 x Amber Vial with Retinoic Acid 0.50 mL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BPE, Triiodothyronine, Insulin, hEGF, Hydrocortisone, Transferrin, Epinephrine, GA-1000, Retinoic Acid
Epithelial Cells
Supplements and growth factors
BPE: 2.0 mL, Triiodothyronine: 0.5 mL, Insulin: 0.5 mL, hEGF: 0.5 mL, Hydrocortisone: 0.5 mL, Transferrin: 0.5 mL, Epinephrine: 0.5 mL, GA-1000: 0.5 mL, Retinoic Acid: 0.5 mL