missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Premier Half-Mask Air Purifying Respirator (APR)

Premier Half-Mask APR features a silicone facepiece with soft, rounded edges and a low-profile nosepiece for low breathing resistance and eyewear compatibility. The twin filters use a ‘click-fit’ system and the four-point harness with neck strap offers improved comfort.

66.00 € - 92.00 €

Specifications

Certifications (EN) EN 140
Color Navy Blue
Design Type Low Profile
Disposable Reusable
Filter Type Twin Filter
View More Specs

Alert:

For use only with certified T-Series cartridges (standards: EN 141, EN 143). The oxygen concentration must be superior to 17%.


Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
11807171
View accessory products View Documents
Honeywell™
1001575
Medium
92.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
12652653
View Documents
Honeywell™
1001576
Large
66.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Material (Facepiece): Silicone
  • Filter: Twin Filter System with Click-Fit Connection
  • Harness Type: Four-Point with Neck Strap
  • Standard/Compliance: EN 140
  • Use with (Application): Laboratories, Pharmaceutical Industry, Oil and Gas Industries
Specifications

Specifications

EN 140
Low Profile
Twin Filter
Half-Mask Air Purifying Respirator (APR)
Laboratories, Pharmaceutical Industry, Oil and Gas Industries
Four-Point with Neck Strap
1 Ea.
Navy Blue
Reusable
Silicone
Silicone
Honeywell™ Click-Fit Filters
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Honeywell™ Premier Half-Mask Air Purifying Respirator (APR)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.