Learn More
Ark Plas Products™ Premium Grade Natural Kynar Barbed Threaded Connectors
Product Code. 11881150
Click to view available options
Port Size:
1/16 in. Barb
Unit Size:
Pack of 10
Product Code. 11881150
Brand: Ark Plas Products™ MEX45DF0
Only null left
Add to basket
Edge
Add to basket
Threaded barbed fittings for forming connections in the laboratory, with a pressure rating of up to 125 P.S.I. without clamps.
  • Molded from virgin, lot-traceable material only
  • Manufactured to the most exacting standards
  • Made with critical use applications in mind
Specifications
Material Kynar PVDF
Packaging Quantity 10 Pack
Port Size 1/16 in. Barb
Thread Size 1/8-27 MNPT
Color Natural
Product Type Barbed Threaded Connector