Learn More
Applied Biosystems™ PrepSEQ™ 1-2-3 Mycoplasma Nucleic Acid Extraction Kit
The Applied Biosystems PrepSEQ 1-2-3 Nucleic Acid Extraction Kit rapidly extracts Mycoplasma genomic DNA from 100 μL sample volumes of research cell lines, stem cell lines, and cell and tissue therapy samples.
Brand:  Applied Biosystems™ 4443789
Product Code. 30000684
  • 1135.00 € / 100 reactions
Explore more special offers
Description
Description
The Applied Biosystems PrepSEQ 1-2-3 Nucleic Acid Extraction Kit rapidly extracts Mycoplasma genomic DNA from 100 μL sample volumes of research cell lines, stem cell lines, and cell and tissue therapy samples. In the PrepSEQ 1-2-3 kit protocol, a rapid procedure recovers DNA from total lysate of the test sample using magnetic-bead based technology. The PrepSEQ 1-2-3 kit is optimized for use with the MycoSEQ Mycoplasma Detection Kit.
Features of the PrepSEQ 1-2-3 Nucleic Acid Extraction Kit include:
  • High recovery of Mycoplasma genomic DNA from mammalian cell culture samples
  • Rapid easy-to-use protocol with results in under 2 hours
  • Compatible with the MycoSEQ Mycoplasma Detection assay
  • Protocol available for automated sample preparation
High DNA recovery sample preparation
Optimized for use with the MycoSEQ Mycoplasma Detection Kit, the PrepSEQ 1-2-3 Nucleic Acid Extraction Kit uses proprietary magnetic bead-based separation technology to extract Mycoplasma DNA from mammalian cell lines. The PrepSEQ 1-2-3 kit extracts Mycoplasma genomic DNA from research cell lines, stem cell lines, and cellular and tissue therapy samples with high efficiency.
The protocol is recommended for use with a starting sample volume of 100 μL with a maximum cell density of 106 cells. The PrepSEQ 1-2-3 kit provides an easy to use procedure consisting of 1) RNase treatment, 2) proteinase treatment, and 3) DNA extraction using the magnetic bead technology (figure below).
The kit isolates Mycoplasma genomic DNA with high sensitivity from total lysate of test sample. The PrepSEQ 1-2-3 kit and the MycoSEQ kit allow routine screening of high-value cell lines using an easy, rapid and sensitive protocol. The figure below demonstrates how a sample that was spiked with low levels of Mycoplasma bovoculi DNA was found negative for Mycoplasma contamination using a commercially prepared kit. When using the PrepSEQ 1-2-3 kit and the MycoSeq kit, the correct positive call for Mycoplasma contamination was made.
Specifications
Specifications
Includes buffers, magnetic beads and enzymes for DNA extraction for Mycoplasma DNA extraction
Mini
Genomic DNA
100 Reactions
100 reactions
Mycoplasma spp.
Magnetic Bead
Cell Culture Supernatants
High-throughput Compatible
PrepSEQ™
100 μL
2 hr.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ PrepSEQ™ 1-2-3 Mycoplasma Nucleic Acid Extraction Kit > 100 Reactions