missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepSEQ™ 1-2-3 Nucleic Acid Extraction Kit

The PrepSEQ 1-2-3 Nucleic Acid Extraction Kit uses magnetic particle-based separation technology to extract DNA from Mycoplasma cells or Mouse Minute Virus (MMV) isolated from a variety of starting material.

Brand:  Applied Biosystems™ 4452222

Product Code. 30000691

  • 1165.00 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The PrepSEQ 1-2-3 Nucleic Acid Extraction Kit uses magnetic particle-based separation technology to extract DNA from Mycoplasma cells or Mouse Minute Virus (MMV) isolated from a variety of starting material, such as infected cell cultures or Mycoplasma liquid cultures.

Using this kit, lysate samples are prepared prior to extracting the target DNA. For samples that contain Mycoplasma, the lysates contain Mycoplasma DNA from the free organisms in the culture media and from organisms released from the infected host cells.

Specifications

Specifications

• PrepSEQ Proteinase K (store at -15°C to -25°C)
• RNASE Cocktail (-15°C to -25°C)
• PrepSEQ Extraction Kit including lysis buffer, Proteinase K buffer, binding solution, wash and elution buffers (room temperature)
• PrepSEQ Magnetic Beads (room temperature)

Magnetic Bead
Genomic DNA, Viral DNA, Bacterial DNA
PrepSEQ™
Genomic DNA
Kit
Cell Culture Supernatants
Cell Culture Contamination Detection
100 Preps
Mouse Minute Virus (MMV), Mycoplasma spp., Vesivirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ PrepSEQ™ 1-2-3 Nucleic Acid Extraction Kit > 100 Preps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.