Learn More
JULABO PRESTO Process Systems
GREENER_CHOICE
The PRESTO systems are designed for precise temperature control as well as rapid temperature changes, making them ideal for reactor vessels, material stress tests, or temperature simulations.
26835.00 € - 39735.00 €
Specifications
Type Process System
View More Specs
Products 4
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
17714869
GREENER_CHOICE
View Documents
JULABO
9420381.N1
A38
26835.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
17724869
GREENER_CHOICE
View Documents
JULABO
9420411.N1
A41
27640.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
17734869
GREENER_CHOICE
View Documents
JULABO
9420701.N1
A70
39735.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
17744869
GREENER_CHOICE
View Documents
JULABO
9421411.N1
W41
28795.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
Description
Description
  • Wide working temperature ranges without changing fluids
  • Ethernet interface
  • SD-Card slot
  • pace optimized design creates more room directly next to the units
  • Connections for USB, Ethernet, RS232, and Alarm output
Specifications
Specifications
Process System