Learn More
Invitrogen™ PRIMER, 5'ALEXA 647 50 NMOL
Custom Primer
Brand:  Invitrogen™ 10660005
130.00 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Product Code. 10434172
  • 195.00 € / Each
Explore more special offers
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Normal or Reversed-phase Chromatography, HPLC or PAGE
Custom 5'Alexa 647 Primer
HPLC
Custom 5'Alexa 647 Primer, 50 nmole, Purified by Normal or Reversed-phase Chromatography, HPLC or PAGE, Used to Design a Custom DNA Oligonucleotide, With any of a Variety of 3' and 5' Modification
50 nmol
Design a Custom DNA Oligonucleotide, with any of a variety of 3' and 5' Modification