Learn More
Invitrogen™ PRIMER 5'HEX 25 NMOL
Custom Primer
Brand:  Invitrogen™ 11862091
56.06 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 10134372
  • 86.25 € / Each
Explore more special offers
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5' HEX Primer, 25 nmol
5' HEX Primer
5' Labeled
25 nmol