Learn More
Invitrogen™ PRIMER INOSINE 200 NMOL
Custom Primer
Brand:  Invitrogen™ 10624039
13.30 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 10515543
  • 17.73 € / Each
Explore more special offers
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Inosine Primer, 200 nmol
Inosine Primer
200 nmol