Learn More
Invitrogen™ PRIMER S-OLIGO 10 UMOL
Custom Primer
Brand: Invitrogen™ 10917045
35.66 EUR valid until 2024-12-14
Use promo code "21632" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
S-Oligo Primer, 10μmol | |
S-Oligo Primer | |
10 μmol |