Learn More
Zeiss™ Primostar 3 - Fixed Kohler Binocular
Robust and compact upright microscope for digital teaching and routine lab work. Configured for transmitted brightfield microscopy with a range of accessories to expand the microscopy techniques available.
Brand:  Zeiss™ 415501-0001-000
1295.45 EUR valid until 2024-11-29
Use promo code "21706" to get your promotional price.
Product Code. 17362800
  • 1685.00 € / 1 set
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The Primostar 3 is a robust yet compact upright microscope for digital teaching and routine lab work.
  • Reliable and robust due to a study metal frame, whilst being light weight.
  • The Fixed-Köhler versions of Primostar 3 come pre-adjusted with a field of view of 20 mm. Selected objective lenses and eyepieces are already in place meaning no time is wasted in the set up.
  • Full-Köhler versions of Primostar 3 with field of view 22 mm give freedom to teach while using Köhler illumination. In pre-defined packages, a selection of objective lenses and eyepieces are already in place. Benefit from a relaxed posture for day-long work with enlarged stage drive.
  • Use the ZEISS Labscope app to connect all your students' microscopes plus WiFi cameras and create a collective learning experience
  • Use Labscope, the imaging app from ZEISS, to display all the live images from your connected microscopes
  • ZEN compatible .czi file format which includes all metadata and a separate annotation layer.
Specifications
Specifications
ABBE 0.9
Binocular Tube