missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Printers for CryoMed™ and CryoPlus™ Freezers

Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with this optional printer.

Brand:  Thermo Scientific™ 4000665

 View more versions of this product

Product Code. 11721579

  • 2904.45 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with Thermo Scientific™ Thermal Printers for CryoMed and CryoPlus™ Freezers. These printers are available in 120V or 220V models, and include cable assembly and RS-232 connector kit.

  • Model 4000565 - 120V, 60Hz printer with cable assembly and RS-232 connector kit
  • Model 4000665 - 220V, 50Hz printer with cable assembly and RS-232 connector kit
  • Model 4000566 - Thermal printer paper (5 rolls/package)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Thermal Data Printer, 220V
All CryoMed Controlled-Rate Freezers
120/220 V
Printer
Warranty varies by country, please contact us for details
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.