Learn More
Epredia™ PrintMate™ Embedding Cassettes
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Brand:  Epredia™ A84210048
 View more versions of this product
Product Code. 15471820
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Designed for performance and user convenience, PrintMate Embedding Cassettes have been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems.  The even, textured printing face provides the optimal surface for high-quality printing every time.  For additional information, the right and left sides are textured for clear handwriting with suitable pens.  The lid catches have been adjusted for ease of use with low opening and closing force, while still giving the security needed while processing.
Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.
PrintMate Embedding Cassettes are also available without lids, for economical use with reuseable metal lids.
Specifications
Specifications
Round Pore
Yes
White
Embedding Biopsy Cassette