Learn More
Epredia™ PrintMate™ Slotted Cassettes
Shop All Epredia Products
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Pack of 750
Quantity:
10 x 75 cassettes (750)/cs
4 x 250 cassettes (1000)/cs
Description
Designed for performance and user convenience, PrintMate Slotted Cassettes have been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low opening and closing force, while still giving the security needed while processing.
Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.
Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.
Specifications
Specifications
| Pore Style | Slotted Pore |
| PrintMate Compatible | Yes |
| Color | Lilac |
| Product Type | Biopsy Cassette |