missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pro-Detect™ Rapid Streptag II Competitive Assay Kit

Pro-Detect™ Rapid Competitive Assay Kit is a single-use, dipstick lateral flow kit for quick, easy detection of tagged proteins in protein expression and purification systems 

Brand:  Thermo Scientific™ A38510

86.01 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 15980513

  • 141.00 € / Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: 10 assays
This item is not returnable. View return policy

Description

Description

  • Lateral flow assay uses membrane-based strips coated with dye-bound conjugated antibodies specific to the tag to provide a qualitative detection within ten minutes
  • Detect tagged proteins directly from cell culture media (supernatant) or cell lysates without any special instrumentation or handling
  • Lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
  • Dilute sample using provided diluent and dip in the lateral flow strip for visual results within 5 to 10 minutes
  • Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentrations
  • Specific—assay does not cross-react with common lysis and protein preparation reagents
  • Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
  • Long shelf life—stable for at least one year at 4°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 10 Rapid Assay Strips
• Sample Diluent, 15 mL

Sufficient for 10 assay determinations

Store at 2–8°C.
Lateral Flow Rapid Assay
Pro-Detect™
Protein tag
Lateral Flow Rapid Assay
Wet Ice
Streptag II-tagged proteins
Chromogenic
Assay
10 Assays
10 Assays
Protein
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.