Learn More
Invitrogen™ ProcartaPlex™ NHP Control Set, 37plex
The ProcartaPlex Non-Human Primate Control Set 37plex contains a high and low control for all analytes included in the Non-Human Primate Cytokine/Chemokine/Growth Factor 37-Plex ProcartaPlex Panel.
Brand:  Invitrogen™ EPX370-40045-CTR
Product Code. 17108905
  • 228.00 € / 1 set
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The ProcartaPlex Non-Human Primate Control Set 37plex contains a high and low control for all analytes included in the Non-Human Primate Cytokine/Chemokine/Growth Factor 37-Plex ProcartaPlex Panel. For a list of analytes included in this set, please refer to the TDS in the Documents section.
Reported application
  • Multiplex Immunoassay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Multiplex Control Set
Non-Human Primate
ProcartaPlex™
Wet Ice
1 set
Luminex™ Instruments
• 1 vial NHP Control High A
• 1 vial NHP Control High B
• 1 vial NHP Control Low A
• 1 vial NHP Control Low B
Store at 2°C to 8°C.