missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Wash Buffer (10X)

ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates.

288.00 €

Specifications

Product Type Buffer
For Use With (Equipment) Luminex™ Instruments
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16396622
View Documents
Invitrogen™
EPX-66666-001
200 mL
288.00 €
200mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates. It is a simple mix of buffers, salts, and mild detergent.

ProcartaPlex Wash Buffer (10X) is included in our basic kits and off-the-shelf panels. Additionally, we offer it here as a stand-alone item.

Reported application

  • Multiplex immunoassay
Specifications

Specifications

Buffer
Luminex™ Instruments
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ ProcartaPlex™ Wash Buffer (10X)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.