Learn More
Corning™ ProCulture™ Fittings
Dual angled sidearm fittings can be used for aseptically transferring medium into or out of angled sidearm spinner flasks or for sparging the medium with gases.
Brand:  Corning™ 4519-168
 View more versions of this product
Product Code. 10767424
  • 580.84 € / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used to secure pH, O2­, or temperature sensors in large spinner flasks with vertical sidearms
  • Fittings are comprised of a PET insert with a fluorocarbon O-Ring and a polypropylene sealing cap.
  • The 316 sensors are held in place by modified polyphenylene oxide thermoplastic nuts with integrated ferrules.
  • The fittings are completely autoclavable.
Spare parts for sidearm fittings, see Cat. Nos. 1395-32LTC, 1395-45LTC, 1395-LTR, and 1395LTMC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3 Tubes, Angled
Stainless Steel
6 L Sidearm Spinner Flask
Each
Autoclavable
3.17 mm
Used to sparge the cell culture medium with gases to enhance cell production.Â
Insert, O-ring, sealing cap
Triple Sidearm
Non-sterile