Learn More
Corning™ ProCulture™ Fittings
Dual angled sidearm fittings can be used for aseptically transferring medium into or out of angled sidearm spinner flasks or for sparging the medium with gases.
Brand:  Corning™ 4519-171
 View more versions of this product
Product Code. 10777614
  • 629.91 € / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used to aseptically transfer medium, sparge the cell culture medium directly or add gases to the head space above the cell culture medium
  • Fittings are comprised of a PET insert with a fluorocarbon O-Ring and a polypropylene sealing cap.
  • One or two 316 stainless steel tubes extend to the bottom of the flask; the other is a shorter 6 length.
  • Both tubes are held in place by modified polyphenylene oxide thermoplastic nuts with integrated ferrules.
  • The fittings are completely autoclavable
Spare parts for sidearm fittings, see Cat. Nos. 1395-32LTC, 1395-45LTC, 1395-LTR, and 1395LTMC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 Tubes, Angled
Stainless Steel
15 L Sidearm Spinner Flask
Each
Autoclavable
6.35 mm
Used to sparge the cell culture medium with gases to enhance cell production.Â
Insert, O-ring, sealing cap
Sidearm
Non-sterile