Learn More
GeneBLAzer™ P2RY11-NFAT-bla CHO-K1 Cells
GeneBLAzer™ P2RY11-NFAT-bla CHO-K1 Cells contain the human Purinergic receptor P2, G protein-coupled, 11 (P2RY11).
Brand:  GeneBLAzer™ K1729
Additional Details : Weight : 0.01000kg
Product Code. 10237953
  • 16120.00 € / Each
Estimated Shipment: 24-07-2024
to see stock.
Add to basket
Description
Description
  • Gene Symbol: P2RY11
  • Accession Number: NM_002566.4
  • Population Status: Single Cell Clone Origin
  • GeneBLAzer™ P2RY11-NFAT-bla CHO-K1 cells contain the human Purinergic receptor P2, G protein-coupled, 11 (P2RY11) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. no. K1534) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.
  • The GeneBLAzer™ P2RY11-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50concentrations of ATP. In addition, GeneBLAzer™ P2RY11-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
  • P2RY11 expression is broadly distributed and has been found in the brain, pituitary, lymphocytes, spleen, intestines, macrophages, lung, stomach, adipose tissue, pancreas, kidney, prostate, heart, placenta, liver, and skeletal muscle. Differentiation of human promyelocytic HL60 cells and maturation of monocyte-derived dendritic cells have been shown to be mediated by ATP activation of P2RY11.
Specifications
Specifications
Liquid Nitrogen
GPCRs
2 x 106 Cells
Fluorescent
P2RY11-NFAT-bla CHO-K1 cells, P2RY11-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis
Dividing Cells