Learn More
GeneBLAzer™ EDG7-NFAT-bla HEK 293T Cells
Brand:  Invitrogen K1511
Additional Details : Weight : 0.01000kg
Product Code. 10267413
  • 16120.00 € / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Specifications
NM_012152.1
Dividing Cells
Fluorescent
EDG7
GeneBLAzer™
EDG7
GeneBLAzer™
Cell-based beta lactamase reporter gene
Contents:
K1511 GeneBLAzer™ EDG7-NFAT-bla HEK293T cells
(LiveBLAzer™-FRET B⁄G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ EDG7-NFAT-bla HEK293T cells
• GeneBLAzer™ EDG7-NFAT-bla HEK293T Assay Protocol
• Certificate of Analysis
Storage:
The GeneBLAzer™ EDG7-NFAT-bla HEK293T cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
FRET
Single Cell Clone Origin
BLA (Beta-Lactamase)
293T (HEK)
GPCR