missing translation for 'onlineSavingsMsg'
Learn More

GeneBLAzer™ OPRL1-Gqi5-NFAT-bla FreeStyle™ 293F Cells

GeneBLAzer™ OPRL1-Gqi5-NFAT-bla FreeStyle™ 293F Cells contain the human opioid/opiate receptor-like-1 receptor (OPRL1).

Brand:  GeneBLAzer™ K1714


Additional Details : Weight : 0.01000kg

Product Code. 10702974

  • 16120.00 € / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

  • Gene Symbol: OPRL1
  • Accession Number: NM_000913
  • Population Status: Single Cell Clone Origin
  • Technique: FRET
  • GeneBLAzer™ Gqi5-NFAT-bla FreeStyle™ 293F cells (Cat. no. K1539) contain the chimeric Gqi5 G protein and the beta-lactamase reporter gene under control of the nuclear factor of activated T cells (NFAT) response element.
  • OPRL1-Gqi5-NFAT-bla FreeStyle™ 293F cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time, and functionally validated for Z-factor and EC50 concentrations of orphanin FQ/nociceptin. These data are found in the Validation & Assay Performance Summary.
  • Orphanin FQ/nociceptin and OPRL1 have been implicated in various physiological functions. The most defined role is their involvement in the mediation of pain in the central nervous system and their possible roles in opiate tolerance, opiate dependence and withdrawal, and the ability to cope with stress and anxiety. Through the use of antagonists, antisense oligos, and transgenic knockout mutants, OPRL1 has been implicated in additional systems including learning, memory, and reproduction.
Specifications

Specifications

Shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt
GPCRs
2 x 106 Cells
Fluorescent
GeneBLAzer™ OPRL1-Gqi5-NFAT-bla Freestyle 293F cells, GeneBLAzer™ OPRL1-Gqi5-NFAT-bla Freestyle 293F Assay
Dividing Cells
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers