Learn More
GeneBLAzer™ M5-NFAT-bla CHO-K1 Cells
GeneBLAzer™ M5-NFAT-bla CHO-K1 Cells contain the human Acetylcholine (Muscarinic) Receptor 5 (M5) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.
Brand:  GeneBLAzer™ K1728
Additional Details : Weight : 0.01000kg
Product Code. 10783544
  • 16120.00 € / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
  • GeneBLAzer™ M5-NFAT-bla CHO-K1 cells contain the human Acetylcholine (Muscarinic) Receptor 5 (M5) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. no. K1534) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.
  • The GeneBLAzer™ M5-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50concentrations of Carbachol. In addition, GeneBLAzer™ M5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
  • Muscarinic acetylcholine receptor subtype 5 (M5) has an unknown physiological relevance primarily because of low expression levels and the lack of M5 receptor-selective ligands. However, knockout studies have suggested that M5 is involved in modulation of central dopamine function and the tone of cerebral blood vessels.
Specifications
Specifications
Store in liquid nitrogen immediately upon receipt
Fluorescent
GeneBLAzer™ M5-NFAT-bla CHO-K1 cells, GeneBLAzer™ M5-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis
Dividing Cells
Gene Symbol: CHRM5; Accession Number: BC041805.1
GPCRs
2 x 106 Cells