missing translation for 'onlineSavingsMsg'
Learn More

uvex Profabutyl B-05R Chemical Protection Gloves

Product Code. 17240501
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Constructed of 100% butyl rubber with 0.50 mm bromobutyl coating for chemical protection when working with polar compounds such as esters and ketones.

  • Butyl rubber chemical protection gloves without stockinette
  • Seamlessly coated with 0.50 mm bromobutyl
  • Enhanced grip in damp or wet applications
  • Resistant to polar compounds such as esters, ketones, aldehydes, acids, and alkalis
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory
Coating Material Bromobutyl
Color Black
Material Butyl Rubber
Size 7
EN388 Standard 2010X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ABIKLNOT
For Use With (Application) Anti-Static Areas, Chemical Handling
Gender Unisex
Glove Cuff Style Gauntlet
Series Profabutyl B-05R
Length (Metric) 35 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.